missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Sgf I Genome Qualified (C) DFS Item

$92.00 - $92.00

Specifications

Cut Site GCGAT.CGC CGC.TAGCG
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1% Thesit™, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Units Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Units Price Quantity  
PR-R7103 Promega™
R7103
8 to 12U/μL 250U 250U Each for $92.00
Specifications

Description

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Description & Specifications

Specifications

GCGAT.CGC CGC.TAGCG
7.4
-20°C
37°C
10mM Tris HCl (pH 7.4), 300mM KCl, 0.1% Thesit™, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol