Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Sfi I Blue/White Cloning and Genome Qualified (B) DFS Item

$72.00 - $72.00

Specifications

Concentration 10U/μL
Cut Site GGCCNNNN.NGGCC CCGGN.NNNNCCGG
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 10mM MgCl2, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
View More Specs
Products
Catalog Number Mfr. No. Includes Product Name Quantity Units Price Quantity    

PR-R6391

DFS Item
 
promega™
R6391
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 250U SfiI, 1 x 0.25mL Muti-Core™10X Buffer SfiI 250U 250U Each for $72.00
Description & Specifications

Specifications

Concentration 10U/μL
Cut Site GGCCNNNN.NGGCC CCGGN.NNNNCCGG
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 10mM MgCl2, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.