Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Sfi I Blue/White Cloning and Genome Qualified (B)


Manufacturer: promega™  R6391

 View more versions of this product

Catalog No. PR-R6391

  • / Each
 

Description & Specifications

Specifications

Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 10mM MgCl2, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Format Liquid
Storage Requirements -20°C
Includes 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 250U SfiI, 1 x 0.25mL Muti-Core™10X Buffer
Concentration 10U/μL
pH 7.4
Units 250U
Product Name SfiI
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Cut Site GGCCNNNN.NGGCC CCGGN.NNNNCCGG
Incubator Temperature 37°C
Quantity 250U