Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Sca I (K)


Manufacturer: promega™  R4214

 View more versions of this product

Catalog No. PR-R4214

  • / Each
 

Description & Specifications

Specifications

Concentration 40 to 80U/μL
pH 7.4
Incubator Temperature 37°C
Quantity 5000U
For Use With (Application) Capable of digesting DNA in <15 min.
Format Liquid
Includes 1 x 1mL 10X Buffer K, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U ScaI (HC)
Units 5,000U
Product Name ScaI (HC)
Quantity 5000U
Applications Capable of digesting DNA in <15 min.
Product Name ScaI (HC)
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
Cut Site AGT.ACT TCA.TGA
Format Liquid