Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Sau3A I (B)


Manufacturer: promega™  R6195

 View more versions of this product

Catalog No. PR-R6195

  • / Each
 

Description & Specifications

Specifications

Cut Site .GATC CTAG.
Product Name Sau3AI
Units 500U
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
Format Liquid
Includes 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U Sau3AI, 1 x 0.25mL Muti-Core™10X Buffer
Incubator Temperature 37°C
Concentration 3 to 10U/μL
Quantity 500U
pH 7.4