Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Sau3A I (B) DFS Item

$54.00 - $211.00

Specifications

Concentration 3 to 10U/μL
Cut Site .GATC CTAG.
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Units Price Quantity    

PR-R6191

DFS Item
 
promega™
R6191
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U Sau3AI, 1 x 0.25mL Muti-Core™10X Buffer 100U 100U Each for $54.00

PR-R6195

DFS Item
 
promega™
R6195
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U Sau3AI, 1 x 0.25mL Muti-Core™10X Buffer 500U 500U Each for $211.00
Description & Specifications

Specifications

Concentration 3 to 10U/μL
Cut Site .GATC CTAG.
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.