Please login to your online account to display your discounted pricing

Restriction Enzymes - Sal I Blue/White Cloning and Genome Qualified (D) DFS Item


Manufacturer: promega corportaion  R4054

 View more versions of this product

Catalog No. PR-R4054

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U Sall (HC)
Concentration 40 to 80U/μL
pH 7.3
Cut Site G.TCGAC CAGCT.G
Product Name SalI (HC)
Format Liquid
Quantity 10000U
Incubator Temperature 37°C
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Units 10,000U
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C