Promega™ Restriction Enzymes - Sac I Blue/White Cloning Qualified (J) DFS Item
Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. SacI recognizes the same DNA sequence but cleaves at a different nucleotide than its neoschizomer, EcoICRI. (G AGCT/C C\TCGA G)
Manufacturer:  Promega™ R4064
 View more versions of this product
Catalog No. PR-R4064
Specifications
Specifications
SacI (HC)
40 to 80U/μL
1 x 1mL 10X Buffer J, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U Sac I (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
7.4
Provides a higher level of quality control for Enzymes used in cloning applications
-20°C
Liquid
GAGCT.C C.TCGAG
37°C
5,000U
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Documents