Promega™ Restriction Enzymes - Rsa I (C) DFS Item
Capable of digesting DNA in 15 minutes or less. Available at high concentration, containing 5,000 units of RsaI at a concentration of 40-80u/μL. (GT/AC CA\TG)
Manufacturer:  Promega™ R4374
 View more versions of this product
Catalog No. PR-R4374
Specifications
Specifications
RsaI (HC)
40 to 80U/μL
1 x 1mL 10X Buffer C, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U RsaI (HC)
7.4
Capable of digesting DNA in <15 min.
−30 to −10°C
Liquid
GT.AC CA.TG
37°C
5,000U
10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol