Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Pvu II (B)


Manufacturer: promega™  R6335

 View more versions of this product

Catalog No. PR-R6335

  • / Each
 

Description & Specifications

Specifications

pH 7.4
For Use With (Application) Active and capable of digestion directly in GoTaq™ Green Master Mix
Units 5,000U
Includes 1 x 1mL 10X Buffer B, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U PvuII, 1 x 0.25mL Multi-Core™ 10X Buffer
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Product Name PvuII
Format Liquid
Concentration 8 to 12U/μL
Incubator Temperature 37°C
Quantity 5000U
Cut Site CAG.CTG GTC.GAC
Storage Requirements -20°C