Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Pvu II (B)


Manufacturer: promega™  R6331

 View more versions of this product

Catalog No. PR-R6331

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U PvuII, 1 x 0.25mL Multi-Core™ 10X Buffer
Quantity 1000U
For Use With (Application) Active and capable of digestion directly in GoTaq™ Green Master Mix
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Product Name PvuII
pH 7.4
Format Liquid
Incubator Temperature 37°C
Cut Site CAG.CTG GTC.GAC
Storage Requirements -20°C
Concentration 8 to 12U/μL
Units 1,000U