Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Pvu II (B)

$52.00 - $197.00

Specifications

Concentration 8 to 12U/μL
Cut Site CAG.CTG GTC.GAC
For Use With (Application) Active and capable of digestion directly in GoTaq™ Green Master Mix
Incubator Temperature 37°C
pH 7.4
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Units Price Quantity    

PR-R6331

 
promega™
R6331
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U PvuII, 1 x 0.25mL Multi-Core™ 10X Buffer 1000U 1,000U Each for $52.00

PR-R6335

 
promega™
R6335
1 x 1mL 10X Buffer B, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U PvuII, 1 x 0.25mL Multi-Core™ 10X Buffer 5000U 5,000U Each for $197.00
Description & Specifications

Specifications

Concentration 8 to 12U/μL
Cut Site CAG.CTG GTC.GAC
For Use With (Application) Active and capable of digestion directly in GoTaq™ Green Master Mix
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.