Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Not I Blue/White Cloning and Genome Qualified (D)


Manufacturer: promega™  R4434

 View more versions of this product

Catalog No. PR-R4434

  • / Each
 

Description & Specifications

Specifications

Units 1,000U
Concentration 40 to 80U/μL
Storage Requirements -20°C
Cut Site GC.GGCCGC CGCCGG.CG
Quantity 1000U
Includes 1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U NotI (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
Storage Buffer 10mM Tris HCl (pH 7.4), 500mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.1% Triton™ X-100, 50% Glycerol
pH 7.4
Product Name NotI (HC)
Quantity 1000U
For Use With (Application) Capable of digesting DNA in <15 min.
Format Liquid
Format Liquid
Product Name NotI (HC)
Applications Capable of digesting DNA in <15 min.