missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Restriction Enzymes - Not I Blue/White Cloning and Genome Qualified (D)
Capable of digesting DNA in 15 minutes or less. (GC/GGCC GC CG CCGG\CG)
$87.55 - $332.11
Specifications
Incubator Temperature | 37°C |
---|---|
pH | 7.4 |
For Use With (Application) | Capable of digesting DNA in <15 min. |
Storage Buffer | 10mM Tris HCl (pH 7.4), 500mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.1% Triton™ X-100, 50% Glycerol |
Cut Site | GC.GGCCGC CGCCGG.CG |
Description
- Capable of digesting DNA in 15 minutes or less
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Genome qualified to ensure optimal performance in genomic analysis applications
- High concentration product no longer available
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
37°C | |
Capable of digesting DNA in <15 min. | |
GC.GGCCGC CGCCGG.CG |
7.4 | |
10mM Tris HCl (pH 7.4), 500mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.1% Triton™ X-100, 50% Glycerol | |
Liquid |