Promega Restriction Enzymes - Nhe I Genome Qualified (B) DFS Item
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications. (G/CTAG C C GATC\G)
Manufacturer:  Promega R6505
 View more versions of this product
Catalog No. PR-R6505
Description
Description
  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Genome qualified to ensure optimal performance in genomic analysis applications
Specifications
Specifications
NheI
10U/μL
1 x 1mL 10X Buffer B, 1 x 1250μL Bovine Serum Albumin, Acetylated, 1 x 250U NheI, 1 x 0.25mL Multi-Core™ 10X Buffer
7.4
Capable of digesting DNA in <15 min.
−30 to −10°C
Liquid
G.CTAGC CGATC.G
37°C
1,250U
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol