Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Msp I (B)

$87.00 - $297.00

Specifications

Concentration 10U/μL
Cut Site C.CGG GGC.C
Incubator Temperature 37°C
pH 7.3
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Units Price Quantity    

PR-R6401

 
promega™
R6401
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2000U MspI, 1 x 0.25mL Muti-Core™10X Buffer 2000U 2,000U Each for $87.00

PR-R6405

 
promega™
R6405
1 x 1mL 10X Buffer B, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U MspI, 1 x 0.25mL Muti-Core™10X Buffer 10000U 10,000U Each for $297.00
Description & Specifications

Specifications

Concentration 10U/μL
Cut Site C.CGG GGC.C
Incubator Temperature 37°C
pH 7.3
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.