Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Msp I (B)


Manufacturer: promega™  R6405

 View more versions of this product

Catalog No. PR-R6405

  • / Each
 

Description & Specifications

Specifications

Product Name MspI
Incubator Temperature 37°C
pH 7.3
Includes 1 x 1mL 10X Buffer B, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U MspI, 1 x 0.25mL Muti-Core™10X Buffer
For Use With (Application) MspI and its Isoschizomer HpaII exhibit different methylation sensitivities
Concentration 10U/μL
Quantity 10000U
Units 10,000U
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Cut Site C.CGG GGC.C
Format Liquid
Product Name MspI
Format Liquid
Quantity 10000U
Applications MspI and its Isoschizomer HpaII exhibit different methylation sensitivities