Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Hpa I (J) DFS Item


Manufacturer: promega™  R6301

 View more versions of this product

Catalog No. PR-R6301

  • / Each
 

Description & Specifications

Specifications

Concentration 3 to 10U/μL
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM Sodium Glutamate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Product Name HpaI (J)
Incubator Temperature 37°C
Units 100U
For Use With (Application) Site-specific methylation
Cut Site GTT.AAC CAA.TTG
Format Liquid
Storage Requirements -20°C
Includes 1 x 1mL 10X Buffer J, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U HpaI, 1 x 0.25mL Multi-Core™ 10X Buffer
Quantity 100U