Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Hpa I (J)

$66.00 - $245.00

Specifications

Applications Site-specific methylation
Concentration 3 to 10U/μL
Cut Site GTT.AAC CAA.TTG
Format Liquid
Incubator Temperature 37°C
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Units Price Quantity    

PR-R6301

 
promega™
R6301
1 x 1mL 10X Buffer J, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U HpaI, 1 x 0.25mL Multi-Core™ 10X Buffer 100U 100U Each for $66.00

PR-R6305

 
promega™
R6305
1 x 1mL 10X Buffer J, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U HpaI, 1 x 0.25mL Multi-Core™ 10X Buffer 500U 500U Each for $245.00
Description & Specifications

Specifications

Applications Site-specific methylation
Concentration 3 to 10U/μL
Cut Site GTT.AAC CAA.TTG
Format Liquid
Incubator Temperature 37°C
Product Name HpaI (J)
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM Sodium Glutamate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.