Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Hpa I (J)


Manufacturer: promega™  R6305

 View more versions of this product

Catalog No. PR-R6305

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM Sodium Glutamate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
For Use With (Application) Site-specific methylation
Quantity 500U
Incubator Temperature 37°C
Product Name HpaI (J)
Includes 1 x 1mL 10X Buffer J, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U HpaI, 1 x 0.25mL Multi-Core™ 10X Buffer
Concentration 3 to 10U/μL
Cut Site GTT.AAC CAA.TTG
Units 500U
Format Liquid