Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Hind III Blue/White Cloning Qualified (E)


Manufacturer: promega™  R4047

 View more versions of this product

Catalog No. PR-R4047

  • / Each
 

Description & Specifications

Specifications

Incubator Temperature 37°C
Units 50,000U
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Concentration 40 to 80U/μL
Cut Site A.AGCTT TTCGA.A
Storage Buffer 10mM Tris HCl (pH 7.4), 250mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Format Liquid
Includes 1 x 1mL 10X Buffer E, 1 x 1250μL Bovine Serum Albumin, Acetylated, 1 x 50000 HindIII, 1 x 1mL Multi-Core™ 10X Buffer
Product Name HindIII
Quantity 50000U
Storage Requirements -20°C
Format Liquid
Product Name HindIII
Quantity 50000U
Applications Provides a higher level of quality control for Enzymes used in cloning applications