missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Restriction Enzymes - Hinf I
Used in gene cloning and protein production experiments. (G/ANT C C TNA\G)
Supplier: Promega R6205
Description
- Format: Liquid
Specifications
10 U/μL | |
37°C | |
10mM Tris HCl (pH 7.3), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
5000 U | |
HinfI |
1 x 1mL 10X Buffer C, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 50000U HinfI, 1 x 0.25mL Multi-Core™ 10X Buffer | |
Site-specific methylation | |
-30°C to -10°C | |
G.ANTC CTNA.G | |
Liquid |