missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Restriction Enzymes - Hinf I
Used in gene cloning and protein production experiments. (G/ANT C C TNA\G)
$63.76 - $259.79
Specifications
Concentration | 10 U/μL |
---|---|
Incubator Temperature | 37°C |
For Use With (Application) | Site-specific methylation |
Storage Buffer | 10mM Tris HCl (pH 7.3), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
Content And Storage | -30°C to -10°C |
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity | ||||
PR-R6201
|
Promega
R6201 |
1000 U | 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 10000U HinfI, 1 x 0.25mL Multi-Core™ 10X Buffer |
N/A
|
|
||||
PR-R6205
|
Promega
R6205 |
5000 U | 1 x 1mL 10X Buffer C, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 50000U HinfI, 1 x 0.25mL Multi-Core™ 10X Buffer |
N/A
|
|
||||
Description
- Format: Liquid
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
10 U/μL | |
Site-specific methylation | |
-30°C to -10°C | |
HinfI |
37°C | |
10mM Tris HCl (pH 7.3), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
G.ANTC CTNA.G | |
Liquid |