Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Hae III (C)


Manufacturer: promega™  R6171

 View more versions of this product

Catalog No. PR-R6171

  • / Each
 

Description & Specifications

Specifications

Incubator Temperature 37°C
Format Liquid
Cut Site GG;CCCC;GG
Product Name HaeIII (C)
Quantity 2500U
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Format Liquid
Includes 1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U HaeIII, 1 x 0.25mL Multi-Core™ 10X Buffer
Units 2,500U
Concentration 10U/μL
Storage Requirements -20°C
Product Name HaeIII (C)
Quantity 2500U
Applications Capable of digesting DNA in <15 min.