Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - EcoR V Blue/White Cloning Qualified (D)


Manufacturer: promega™  R4354

 View more versions of this product

Catalog No. PR-R4354

  • / Each
 

Description & Specifications

Specifications

Product Name EcoRV (HC)
Incubator Temperature 37°C
pH 7.4
Includes 1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U EcoRV (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
Quantity 10,000U
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Cut Site GAT.ATC
Storage Requirements -20°C
Concentration 40 to 80U/μL
Format Liquid
Product Name EcoRV (HC)
Quantity 10,000U
Applications Capable of digesting DNA in <15 min.
Format Liquid