Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - EcoR I Blue/White Cloning Qualified (H)


Manufacturer: promega™  R4017

 View more versions of this product

Catalog No. PR-R4017

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 1mL 10X Buffer H, 1 x 1250μL Bovine Serum Albumin, Acetylated, 1 x 50000U EcoRI (HC)
Quantity 50,000U
For Use With (Application) Capable of digesting DNA in <15 min.
Format Liquid
Cut Site G.AATTC
Incubator Temperature 37°C
Storage Requirements -20°C
Format Liquid
pH 7.4
Product Name EcoRI (HC)
Concentration 40 to 80U/μL
Storage Buffer 10mM Tris HCl (pH 7.4), 400mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol
Product Name EcoRI (HC)
Quantity 50,000U
Applications Capable of digesting DNA in <15 min.