Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Dde I (D)


Manufacturer: promega™  R6291

 View more versions of this product

Catalog No. PR-R6291

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 200U DdeI, 1 x 0.25mL Multi-Core™ 10X Buffer
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Requirements -20°C
Format Liquid
Cut Site C.TNAG
Product Name Dde I
Quantity 200U
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.01% Triton™ X-100, 50% Glycerol
Incubator Temperature 37°C
Applications Capable of digesting DNA in <15 min.
pH 7.9
Concentration 10U/μL
Product Name Dde I
Quantity 200U
Format Liquid