Promega™ Restriction Enzymes - Dde I (D) DFS Item
Capable of digesting DNA in 15 minutes or less. DdeI produces an ambiguous extension, which is difficult to ligate. (C/TNA G G ANT\C)
Manufacturer:  Promega™ R6295
 View more versions of this product
Catalog No. PR-R6295
Specifications
Specifications
DdeI
10U/μL
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U DdeI, 1 x 0.25mL Multi-Core™ 10X Buffer
7.9
Capable of digesting DNA in <15 min.
−30 to −10°C
Liquid
C.TNAG
37°C
1000U
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.01% Triton™ X-100, 50% Glycerol