Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Cla I Blue/White Cloning and Genome Qualified (C)

$87.00 - $289.00

Specifications

Applications Capable of digesting DNA in <15 min.
Concentration 10U/μL
Cut Site AT.CGAT
Format Liquid
Incubator Temperature 37°C
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

PR-R6551

 
promega™
R6551
1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U ClaI, 1 x 0.25mL Multi-Core™ 10X Buffer 500U Each for $87.00

PR-R6555

 
promega™
R6555
1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U ClaI, 1 x 0.25mL Multi-Core™ 10X Buffer 2500U Each for $289.00
Description & Specifications

Specifications

Applications Capable of digesting DNA in <15 min.
Concentration 10U/μL
Cut Site AT.CGAT
Format Liquid
Incubator Temperature 37°C
pH 7.9
Product Name Cla I
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM Sodium Glutamate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.