Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Cla I Blue/White Cloning and Genome Qualified (C) DFS Item


Manufacturer: promega™  R6555

 View more versions of this product

Catalog No. PR-R6555

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Concentration 10U/μL
Includes 1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U ClaI, 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM Sodium Glutamate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
For Use With (Application) Capable of digesting DNA in <15 min.
pH 7.9
Storage Requirements -20°C
Cut Site AT.CGAT
Quantity 2500U
Product Name Cla I