Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Bgl II (D) DFS Item


Manufacturer: promega™  R6085

 View more versions of this product

Catalog No. PR-R6085

  • / Each
 

Description & Specifications

Specifications

Concentration 10U/μL
Cut Site A.GATCT TCTAG.A
Incubator Temperature 37°C
Storage Buffer 10mM Tris HCl (pH 7.3), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
For Use With (Application) Ends generated with BglII can be directly ligated to ends generated with BamHI, BclI and XhoII
pH 7.3
Quantity 2,500U
Product Name BglII
Format Liquid
Includes 1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U BglII
Storage Requirements -20°C