Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Bgl II (D)


Manufacturer: promega™  R6081

 View more versions of this product

Catalog No. PR-R6081

  • / Each
 

Description & Specifications

Specifications

Storage Buffer 10mM Tris HCl (pH 7.3), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
Format Liquid
Product Name BglII
Concentration 10U/μL
Quantity 500U
Incubator Temperature 37°C
Includes 1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U BglII
pH 7.3
Cut Site A.GATCT TCTAG.A
For Use With (Application) Ends generated with BglII can be directly ligated to ends generated with BamHI, BclI and XhoII