Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Bgl II (D)

$47.00 - $179.00

Specifications

Concentration 10U/μL
Cut Site A.GATCT TCTAG.A
Incubator Temperature 37°C
pH 7.3
Storage Buffer 10mM Tris HCl (pH 7.3), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

PR-R6085

 
promega™
R6085
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U BglII 2,500U Each for $179.00

PR-R6081

 
promega™
R6081
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U BglII 500U Each for $47.00
Description & Specifications

Specifications

Concentration 10U/μL
Cut Site A.GATCT TCTAG.A
Incubator Temperature 37°C
pH 7.3
Storage Buffer 10mM Tris HCl (pH 7.3), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.