Promega™ Restriction Enzymes - Bgl I Genome Qualified (D) DFS Item
Genome qualified to ensure optimal performance in genomic analysis applications. (GCCN NNN/NGGC CGGN\NNN NCCG)
Manufacturer:  Promega™ R4074
 View more versions of this product
Catalog No. PR-R4074
Specifications
Specifications
BglI (HC)
40 to 80U/μL
1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U BglI (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
7.4
Assayed to ensure optimal performance in genomic analysis applications
-20°C
Liquid
GCCNNNN.NGGC CGGN.NNNNCCG
37°C
5,000U
10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Documents