Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Bgl I Genome Qualified (D)


Manufacturer: promega™  R6077

 View more versions of this product

Catalog No. PR-R6077

  • / Each
 

Description & Specifications

Specifications

Storage Buffer 10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Product Name BglI
Format Liquid
Storage Requirements -20°C
Includes 1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 5000U BglI
Cut Site GCCNNNN.NGGC
Incubator Temperature 37°C
For Use With (Application) Assayed to ensure optimal performance in genomic analysis applications
Quantity 5,000U
pH 7.4
Concentration 10U/μL