Promega™ Restriction Enzymes - BamH I Blue/White Cloning Qualified (E) DFS Item
Capable of digesting DNA in 15 minutes or less. Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. (G/GATC C C CTAG\G)
Manufacturer:  Promega™ R6025
 View more versions of this product
Catalog No. PR-R6025
Description
Description
  • Active and capable of digestion directly in GoTaq™ Green Master Mix
  • Ends generated with BamHI can be directly ligated to ends generated with BglII, BclI and XhoII; these restriction sites are not regenerated in the ligation product
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Specifications
BamHI
10U/μL
1 x 1mL 10X Buffer E, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 12500U BamHI, 1 x 0.25mL Multi-Core™ 10X Buffer
7.4
Provides a higher level of quality control for Enzymes used in cloning applications
−30 to −10°C
Liquid
G.GATCC CCTAG.G
37°C
12,500U
10mM Tris HCl (pH 7.4), 300mM KCl, 5mM MgCl2, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol