Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Bal I (G)


Manufacturer: promega™  R6691

 View more versions of this product

Catalog No. PR-R6691

  • / Each
 

Description & Specifications

Specifications

Cut Site TGG.CCA ACC.GGT
Product Name BalI
Storage Requirements -20°C
Quantity 50U
Incubator Temperature 37°C
Includes 1 x 1mL 10X Buffer G, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U BalI
Format Liquid
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Concentration 2 to 10U/μL
Product Name BalI
pH 7.4
Format Liquid
Quantity 50U