Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Bal I (G)


Manufacturer: promega™  R6695

 View more versions of this product

Catalog No. PR-R6695

  • / Each
 

Description & Specifications

Specifications

Incubator Temperature 37°C
pH 7.4
Includes 1 x 1mL 10X Buffer G, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 250U BalI
Quantity 250U
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
Format Liquid
Concentration 2 to 10U/μL
Cut Site TGG.CCA ACC.GGT
Format Liquid
Product Name BalI
Product Name BalI
Quantity 250U