Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Ava II (C) DFS Item


Manufacturer: promega™  R6135

 View more versions of this product

Catalog No. PR-R6135

  • / Each
 

Description & Specifications

Specifications

Product Name AvaII
pH 7.4
Quantity 1000U
Incubator Temperature 37°C
Format Liquid
Includes 1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U AvaII, 1 x 0.25mL Multi-Core™ 10X Buffer
Concentration 1 to 10U/μL
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM KCl, 0.1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
Cut Site G.G(A/T)CC CC(T/A)G.G