Please login to your online account to display your discounted pricing

Promega™ Restriction Enzymes - Ava II (C)

$46.00 - $301.00

Specifications

Concentration 1 to 10U/μL
Cut Site G.G(A/T)CC CC(T/A)G.G
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM KCl, 0.1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

PR-R6135

 
promega™
R6135
1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U AvaII, 1 x 0.25mL Multi-Core™ 10X Buffer 1000U Each for $301.00

PR-R6131

 
promega™
R6131
1 x 1mL 10X Buffer C, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U AvaII, 1 x 0.25mL Multi-Core™ 10X Buffer 100U Each for $46.00
Description & Specifications

Specifications

Concentration 1 to 10U/μL
Cut Site G.G(A/T)CC CC(T/A)G.G
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM KCl, 0.1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
Storage Requirements -20°C

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.