missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega Replacement Products for the Wizard™ Magnetic DNA Purification System for Food DFS Item

Components of the Wizard™ Magnetic DNA Purification System for Food, which is intended for isolating genomic DNA from a variety of food samples including: corn seeds, corn meal, soy beans, soy flour, soy milk, corn oil, soybean oil, canola oil, etc.

Manufacturer:  Promega Z3191

 View more versions of this product

Catalog No. PRZ3191



Description

Description

  • Store at 22° to 25°C
  • Lysis buffer A and B: 100mL, Precipitation solution: 150mL

Specifications

Specifications

Purifies DNA from a variety of food samples including corn seeds, cornmeal, soybeans, Soy flour and Soy milk
Lysis buffer; B
15 to 30°C
Wizard™ Magnetic DNA purification system for food
100mL
Documents
Promotions

Promotions