Please login to your online account to display your discounted pricing

Promega™ ReliaPrep™ FFPE gDNA Miniprep System DFS Item


Isolation of genomic DNA from formalin-fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion.

Manufacturer: promega™  A2351

 View more versions of this product

Catalog No. PRA2351

  • / Each
 

Description & Specifications

Specifications

Component 5mL mineral oil, 2 x 1mL lysis buffer, 250μL proteinase K, 3.25mL BL buffer, 3mL wash solution, 200μL RNase A, 15mL elution buffer, collection tubes (10 tubes/package), ReliaPrep™ FFP
For Use With (Application) Purifying genomic dna from formalin-fixed paraffin-embedded tissue
Product Type Ffpe Gdna Miniprep System
Storage Requirements Room temperature