missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega Maxwell HT 96 gDNA Blood Isolation System DFS Item

The Maxwell HT 96 gDNA Blood Isolation System provides a simple and reliable method for the rapid isolation of gDNA in a multiwell format. The purified gDNA can be used directly in PCR assays, microarrays and next-generation sequencing applications.

$336.17 - $1,176.63

Specifications

Format Multiwell
For Use With (Application) Rapid isolation of genomic DNA
View More Specs

Includes: Binding Buffer, ReliaPrep Resin, Wash Buffer, 10mM EDTA (pH 8.0), 25mM Tris-HCl (pH 8.0), Proteinase K (PK) Solution, RNase A Solution, Cell Lysis Buffer (CLD)

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRA2671 View Documents Promega
A2671
4 x 96 Preps Each for $1,176.63
 
PRA2670 View Documents Promega
A2670
1 x 96 Preps Each for $336.17
 
Description

Description

  • High-Throughput DNA Extraction in 24-or 96-Well Format
  • Robust, precipitation-free protocol, No chance of lost pellets.
  • High yields of pure DNA from pristine and challenged or hemolysed samples.
  • Pure gDNA ready for demanding applications; samples in solution; no resuspension required
Specifications

Specifications

Multiwell
Rapid isolation of genomic DNA