Please login to your online account to display your discounted pricing

Promega™ Random and Oligo (dT) Primers


For first-strand cDNA synthesis and cloning

Manufacturer: promega™  C1181

 View more versions of this product

Catalog No. PR-C1181

  • / Each
 

Description & Specifications

Specifications

Primer Random
For Use With (Application) cDNA synthesis and cloning
Quantity 20μg
Storage Requirements -20°C
Format Liquid
Product Type Random Primer
pH 7