Please login to your online account to display your discounted pricing

Promega™ Random and Oligo (dT) Primers DFS Item


For first-strand cDNA synthesis and cloning

Manufacturer: promega™  C1101

 View more versions of this product

Catalog No. PR-C1101

  • / Each
 

Description & Specifications

Specifications

Primer Oligo (dT)15
Storage Requirements -20°C
pH 7
Quantity 20μg
For Use With (Application) cDNA synthesis with a Reverse Transcriptase
Format Liquid
Product Type Oligo (dT)15 Primer