missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega Solid Standard for QuantiFluor™-ST Fluorometer DFS Item

The QuantiFluor Fluorometers are small, affordable and sensitive instruments, providing easy and accurate fluorescence measurements. Both instruments provide high sensitivity for fluorescence measurements within minutes.

Manufacturer:  Promega E6113

Catalog No. PRE6113



Description

Description

  • For Use With QuantiFluor(R)-ST and QuantiFluor(R)-P Instruments
  • Instruments are discontinued
  • Accessories available until March 1, 2019
Specifications

Specifications

Solid Standard
1/ea
QuantiFluor-ST Fluorometer only
Verify if any instrument settings have been changed
Store at 5 to 40°C
SDS
Documents
Product Certifications
Promotions

Promotions