Please login to your online account to display your discounted pricing

Promega™ PureYield™ RNA Midiprep System


Isolates intact total RNA with no detectable genomic DNA carryover using a novel, non-DNase technique designed for sensitive analysis methods

Manufacturer: promega™  Z3370

 View more versions of this product

Catalog No. PRZ3370

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Enables rapid purification of total RNA
Format Spin or vacuum
Quantity 100 Preps
Quantity 100 Preps
Format Spin or vacuum