Please login to your online account to display your discounted pricing

Promega™ PureYield™ Plasmid Miniprep System


Isolates transfection-quality plasmid DNA directly from culture in as little as 10 minutes

Manufacturer: promega™  A6810

 View more versions of this product

 Disclaimers

U.S. Patent 6,194,562, Australian Patent 740145, Canadian Patent 2,329,067.

Catalog No. PRA6810

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Isolates transfection-quality plasmid DNA directly from culture in as little as 10 min.
Voltage 110V
Includes Cell Lysis Buffer, column wash solution, Elution Buffer, endotoxin removal wash, neutralization solution, PureYield™ collection tubes, pureYield minicolumns
Product Type Start-Up Kit
Quantity Each
Storage Requirements Store at room temperature.
Product Name Start-Up Kit
Quantity Each