Please login to your online account to display your discounted pricing

Promega™ PureYield™ Plasmid Miniprep System


Isolates transfection-quality plasmid DNA directly from culture in as little as 10 minutes

Manufacturer: promega™  A1222

 View more versions of this product

 Disclaimers

U.S. Patent 6,194,562, Australian Patent 740145, Canadian Patent 2,329,067.

Catalog No. PRA1222

  • / Each
 

Description & Specifications

Specifications

Includes 27mL Cell Lysis Buffer, 90mL neutralization solution, 25mL column wash solution, 10mL Elution Buffer, 2 x 27mL endotoxin removal wash, 250 PureYield™ minicolumns, 250 PureYield™ collection tubes
Storage Requirements Store at room temperature.
For Use With (Application) Isolates transfection-quality plasmid DNA directly from culture in as little as 10 min.
Product Type Plasmid Isolation Kit
Quantity 250 Preps