Please login to your online account to display your discounted pricing

Promega™ PureYield™ Plasmid Miniprep System


Isolates transfection-quality plasmid DNA directly from culture in as little as 10 minutes

Manufacturer: promega™  A1223

 View more versions of this product

 Disclaimers

U.S. Patent 6,194,562, Australian Patent 740145, Canadian Patent 2,329,067.

Catalog No. PRA1223

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Isolates transfection-quality plasmid DNA directly from culture in as little as 10 min.
Includes 11mL Cell Lysis Buffer, 36mL neutralization solution, 9mL Column wash solution, 10mL Elution Buffer, 20.5mL endotoxin removal wash, 2 x 250 PureYield™ minicolumns, 2 x 250 PureYield™ collection tubes
Product Type Plasmid Isolation Kit
Quantity 100 Preps
Storage Requirements Store at room temperature.
Quantity 100 Preps
Product Name Plasmid Isolation Kit